MilliporeSigma
Cytology and receptor architecture of human anterior cingulate cortex.
The Journal of comparative neurology (2008-04-12)
Nicola Palomero-Gallagher, Hartmut Mohlberg, Karl Zilles, Brent Vogt
ABSTRACT
Anterior cingulate cortex (ACC) is involved in emotion, mood, and autonomic regulation. Although a subgenual part of ACC (sACC) may be vulnerable in depression and area 25 is cytologically unique, there are no assessments that contrast this region with pregenual ACC (pACC). Thus, we undertook independent multimodal verifications of architectural differences among subregions and areas. Areas 24a and 24b have pregenual and subgenual components. The latter have a thin layer III. Area 24c has dorsal (pd24c) and ventral (pv24c) parts. Area pd24c has larger neurofilament-expressing neurons in layer Va, and neurons in Vb form aggregates in area pv24c. Area pd24c occupies both banks of the cingulate sulcus, with pv24c on the ventral bank. Layer III of pd24cd has many larger neurofilament-expressing neurons and a richer dendritic plexus. Area 32 has pregenual (p32) and subgenual (s32) components. Layer II in s32 is of particular note because it has a neuron-dense IIa and sparse IIb. Area 25 has anterior (25a) and posterior (25p) parts; 25p has the thinnest layer III in the cingulate gyrus. Area 25a contains significantly higher AMPA, kainate, NMDA, GABA(A), GABA(B), and alpha(1) densities than 25p. Area 33 continues around the genu and ventrally to encompass the full caudal extent of area 25. Subgenual ACC has significantly higher GABA(A), GABA(B), benzodiazepine (BZ), alpha(1), and 5-HT(1A) densities than pACC. GABA(B), BZ, and alpha(1) binding confirms the subdivision of area pd24c. In conclusion, ACC comprises two parts that are unique in terms of their cytoarchitecture and neurotransmitter receptor organization.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis