Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0001569

Mesalazine impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Aminophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Beilstein:
606075
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mesalazine, mesalamine

manufacturer/tradename

EDQM

mp

170-175 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccccc1O

InChI

1S/C6H7NO/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2

InChI key

CDAWCLOXVUBKRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mesalazine impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

334.4 °F - closed cup

Flash Point(C)

168 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen Flávia Moreira Gabriel et al.
Electrophoresis, 35(16), 2325-2332 (2014-08-13)
This paper describes the effects of different modes and engraving parameters on the dimensions of microfluidic structures produced in PMMA using laser engraving. The engraving modes included raster and vector, while the explored engraving parameters included power, speed, frequency, resolution
Zhenke Zhu et al.
Journal of hazardous materials, 279, 436-443 (2014-08-06)
The reduction of nitroaromatic compounds (listed as a priority pollutant) in natural subsurface environments typically coexists with dissimilatory reduction of iron oxides effected by dissolved organic matter (DOM). Investigating the impact of the DOM that influences those reduction processes is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service