Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1419

Supelco

Sulfanilic Acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sulfanilic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
908765
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1633506

API family

sulfadimethoxine, sulfadiazine, mesalazine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfanilic Acid is a synthetic organic compound, obtained via sulfonation of aniline with sulfuric acid. It is widely used for the production of optical brightening agents, synthetic organic dyes, food colorants and additives.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3847 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luís Belo et al.
PloS one, 9(6), e98467-e98467 (2014-06-06)
Bilirubin has potential antioxidant and anti-inflammatory properties. The UGT1A1*28 polymorphism (TA repeats in the promoter region) is a major determinant of bilirubin levels and recent evidence suggests that raised adiposity may also be a contributing factor. We aimed to study
Aminul Islam et al.
Journal of AOAC International, 98(1), 165-175 (2015-04-11)
Amberlite® XAD-16 was functionalized with p-aminobenzene sulfonic acid via an azo spacer in order to prepare a new chelating resin, which was then characterized by water regain value, hydrogen ion capacity, elemental analyses, and IR spectral and thermal studies. The
Kraingkrai Ponhong et al.
Talanta, 133, 71-76 (2014-12-02)
A novel four-channel simultaneous injection effective mixing flow analysis (SIEMA) system has been assembled for successive determination of bilirubin and creatinine in urinary samples. The chemical variables and physical parameters in the flow system were optimized for the enhancement of
Virginia A Pereira et al.
Malaria journal, 14, 30-30 (2015-01-30)
Cytokines play an important role in human immune responses to malaria and variation in their production may influence the course of infection and determine the outcome of the disease. The differential production of cytokines has been linked to single nucleotide
Xiaojun Zhou et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11834-11842 (2014-06-25)
This paper presents a facile method for the fabrication of uniform hollow mesoporous silica nanoparticles (HMSNs) with tunable shell thickness and pore size. In this method, a series of amphiphilic block copolymers of polystyrene-b-poly (acrylic acid) (PS-b-PAA) with different hydrophobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service