MilliporeSigma
  • 2-Nitrophenol reduction promoted by S. putrefaciens 200 and biogenic ferrous iron: the role of different size-fractions of dissolved organic matter.
2-Nitrophenol reduction promoted by S. putrefaciens 200 and biogenic ferrous iron: the role of different size-fractions of dissolved organic matter.
Journal of hazardous materials (2014-08-06)
Zhenke Zhu, Liang Tao, Fangbai Li
ABSTRACT
The reduction of nitroaromatic compounds (listed as a priority pollutant) in natural subsurface environments typically coexists with dissimilatory reduction of iron oxides effected by dissolved organic matter (DOM). Investigating the impact of the DOM that influences those reduction processes is crucial for understanding and predicting the geochemical fate of these environmental species. This study investigated the impact of different molecular weight DOM fractions (DMWDs) on the 2-nitrophenol (2-NP) reduction by S. putrefaciens 200 (SP200) and α-Fe2O3 with lactate (excluding electron donor interference). Kinetic measurements demonstrated that 2-NP reduction rates were affected by the redox reactivity of active species under DMWDs (denoted as L-DOM, M-DOM, and H-DOM). The enhanced reduction rates are consistent with the negative shifts in peak oxidation potential values, the increases in HA-like/FA-like values, aromaticity index values and electron transfer capacity values. L-DOM acted mainly as ligands to complex Fe(II), whereas the significant role of H-DOM in reductive reactions should be acting as an electron shuttle, transferring electrons from SP200 to Fe(III) and 2-NP and from biogenic Fe(II) to 2-NP, further accelerating the 2-NP reductions. Those observations provide valuable insights into the role of DOM in the biogeochemical redox processes and the remediation of contaminated soil in a natural environment.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Aminopurine, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Mesalazine impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C