Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001482

Hydrocortisone acetate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydrocortisone 21-acetate, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O6
CAS Number:
Molecular Weight:
404.50
Beilstein:
2066841
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hydrocortisone

manufacturer/tradename

EDQM

mp

223 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

ALEXXDVDDISNDU-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocortisone acetate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adamo Fini et al.
AAPS PharmSciTech, 9(3), 762-768 (2008-06-20)
The purpose of this research was the preparation of four formulations containing hydrocortisone acetate (HCA) for topical application, including two aqueous systems (hydrophilic microemulsion and aqueous gel) and two systems with dominant hydrophobicity (hydrophobic microemulsion and ointment). The formulations were
Timothy Kassis et al.
PLoS neglected tropical diseases, 8(11), e3305-e3305 (2014-11-21)
Lymphatic Filariasis, a Neglected Tropical Disease, is caused by thread-like parasitic worms, including B. malayi, which migrate to the human lymphatic system following transmission. The parasites reside in collecting lymphatic vessels and lymph nodes for years, often resulting in lymphedema
Janyce A Sugui et al.
Journal of clinical microbiology, 52(10), 3707-3721 (2014-08-08)
Aspergillus section Fumigati contains 12 clinically relevant species. Among these Aspergillus species, A. fumigatus is the most frequent agent of invasive aspergillosis, followed by A. lentulus and A. viridinutans. Genealogical concordance and mating experiments were performed to examine the relationship
Dan-Ni Wang et al.
Medical mycology, 52(5), 506-518 (2014-05-23)
Gliotoxin is an important virulence factor of Aspergillus fumigatus. Although GliA putatively belongs to the major facilitator superfamily in the gliotoxin biosynthesis cluster, its roles remain unclear. To determine the function of GliA, we disrupted gliA in A. fumigatus. gliA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service