Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000577

Pancuronium bromide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pancuronium bromide, 1,1′-[(2β,3α,5α,16β,17β)-3,17-bis(Acetyloxy)androstane-2,16-diyl]bis(1-methylpiperidinium) dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H60Br2N2O4
CAS Number:
Molecular Weight:
732.67
Beilstein:
4226892
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pancuronium 

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].[Br-].[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@]4(C)[C@@H](OC(C)=O)[C@H](C[C@@]34[H])[N+]5(C)CCCCC5)[C@@]1(C)C[C@@H]([C@H](C2)OC(C)=O)[N+]6(C)CCCCC6

InChI

1S/C35H60N2O4.2BrH/c1-24(38)40-32-21-26-13-14-27-28(35(26,4)23-31(32)37(6)19-11-8-12-20-37)15-16-34(3)29(27)22-30(33(34)41-25(2)39)36(5)17-9-7-10-18-36;;/h26-33H,7-23H2,1-6H3;2*1H/q+2;;/p-2/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;;/m0../s1

InChI key

NPIJXCQZLFKBMV-YTGGZNJNSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pancuronium bromide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Roizen et al.
Annals of internal medicine, 88(1), 64-68 (1978-01-01)
Pancuronium bromide is a nondepolarizing muscle relaxant approved to induce skeletal muscle relaxation during anesthesia and to facilitate the management of patients undergoing mechanical ventilation. The use of pancuronium bromide during surgery led to the appreciation that it has advantages
M Bączyk et al.
The Journal of physiology, 592(19), 4313-4328 (2014-08-03)
The main aim of the present study was to examine to what extent long-lasting subcortical actions of transcranial direct current stimulation (tDCS) may be related to its presynaptic actions. This was investigated in the red nucleus, where tDCS was recently
Alina Teliban et al.
Journal of neurophysiology, 111(10), 2071-2083 (2014-02-28)
Intact and injured cutaneous C-fibers in the rat sural nerve are cold sensitive, heat sensitive, and/or mechanosensitive. Cold-sensitive fibers are either low-threshold type 1 cold sensitive or high-threshold type 2 cold sensitive. The hypothesis was tested, in intact and injured
J Simoes et al.
Journal of perinatology : official journal of the California Perinatal Association, 9(2), 215-218 (1989-06-01)
Nursing care of the newborn in respiratory distress has become highly sophisticated and technical, requiring skill in principles of neonatal assessment and therapeutic interventions. It is important to be knowledgeable about the technical aspects of the equipment and monitors. However
Pancuronium bromide: a review of its pharmacological properties and clinical application.
T M Speight et al.
Drugs, 4(3), 163-226 (1972-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service