MilliporeSigma
Nursing interventions: the infant treated with pancuronium bromide.
Journal of perinatology : official journal of the California Perinatal Association (1989-06-01)
J Simoes, C Gagnon
ABSTRACT
Nursing care of the newborn in respiratory distress has become highly sophisticated and technical, requiring skill in principles of neonatal assessment and therapeutic interventions. It is important to be knowledgeable about the technical aspects of the equipment and monitors. However, focus should always remain on the infant as an individual within a family unit.
Product Description
Pancuronium bromide, European Pharmacopoeia (EP) Reference Standard
Pancuronium bromide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pancuronium bromide