Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W233218

Sigma-Aldrich

Cognac oil

white, natural, FG

Synonym(s):

Vitis vinifera, Grape seed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2332
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
fruity; wine-like
grade:
FG
Fragrance grade
natural
biological source:
Vitis vinifera
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

Vitis vinifera

Quality Level

grade

FG
Fragrance grade
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.50

refractive index

n20/D 1.428

density

0.870 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; wine-like

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.1 °F - closed cup

Flash Point(C)

66.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service