Skip to Content
MilliporeSigma
All Photos(1)

Documents

T5550

Supelco

Tea extract from Camellia sinensis

analytical standard, ≥80% theaflavins (theaflavin and theaflavin gallates) basis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥80% theaflavins (theaflavin and theaflavin gallates) basis

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

April N Ilacqua et al.
Biochemical and biophysical research communications, 491(2), 449-454 (2017-07-18)
Theaflavins, flavonoids found in black tea, exhibit a variety of health-promoting activities, but the mechanisms by which they act are not clear. Here, we assess the effects of black tea extract and isolated theaflavins on Dictyostelium discoideum, a model organism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service