Skip to Content
MilliporeSigma
All Photos(1)

Documents

P1204

Sigma-Aldrich

Polyphenon 60 from green tea

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

Other Notes

Extract containing mixture of polyphenolic compounds

Quality

Minimum 60% total catechins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina M Noritake et al.
BMC complementary and alternative medicine, 17(1), 461-461 (2017-09-15)
Plants produce secondary metabolites that often possess widespread bioactivity, and are then known as phytochemicals. We previously determined that several phytochemical-rich food-derived preparations were active against pathogenic foodborne bacteria. Trichomonads produce disease (trichomoniasis) in humans and in certain animals. Trichomonads
Yu-Jin Kim et al.
International journal of molecular sciences, 21(14) (2020-07-12)
Although cell wall dynamics, particularly modification of homogalacturonan (HGA, a major component of pectin) during pollen tube growth, have been extensively studied in dicot plants, little is known about how modification of the pollen tube cell wall regulates growth in
Olivier Habrylo et al.
International journal of biological macromolecules, 112, 499-508 (2018-02-01)
Pectinases represent about one fifth of the enzyme worldwide market due their wide range of biotechnological applications. Current commercial pectinases are exclusively obtained from microbial sources, but here we report a pectin methylesterase (Sl-PME) and an endo-polygalacturonase (Sl-EPG) bioprospected from
Mélanie L'Enfant et al.
International journal of biological macromolecules, 81, 681-691 (2015-09-08)
Pectin methylesterases (PMEs) play a central role in pectin remodeling during plant development. They are also present in phytopathogens such as bacteria and fungi. We investigated the substrate specificity and pH dependence of plant and fungi PMEs using tailor-made pectic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service