Skip to Content
MilliporeSigma
All Photos(2)

Documents

S0252

Sigma-Aldrich

Sulfobromophthalein disodium salt hydrate

used to study hepatocyte transport functions

Synonym(s):

BSP, Bromsulfalein sodium disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H8Br4Na2O10S2 · xH2O
CAS Number:
Molecular Weight:
838.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

form

powder

composition

Water (by Karl Fischer): ≤ 8%

Sodium (Na) anhydrous: 4.6-5.8%

color

white to light gray

solubility

water: 50 mg/mL, clear to hazy

λmax

575-581 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[H]O[H].Oc1ccc(cc1S([O-])(=O)=O)C2(OC(=O)c3c(Br)c(Br)c(Br)c(Br)c23)c4ccc(O)c(c4)S([O-])(=O)=O

InChI

1S/C20H10Br4O10S2.2Na.H2O/c21-15-13-14(16(22)18(24)17(15)23)20(34-19(13)27,7-1-3-9(25)11(5-7)35(28,29)30)8-2-4-10(26)12(6-8)36(31,32)33;;;/h1-6,25-26H,(H,28,29,30)(H,31,32,33);;;1H2/q;2*+1;/p-2

InChI key

CDTNUNQQEUGDSD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sulfobromophthalein disodium salt hydrate has been used as a positive control to study the interaction of 11-keto-β-boswellic acid (KBA) and 3-acetyl-11-keto-β-boswellic acid (AKBA) with organic anion transporter OATP1B3.

Biochem/physiol Actions

Sulfobromophthalein (BSP) is a high affinity organic ion substrate for organic anion transporting polypeptides (OATPs) and has been used as a substrate for multidrug resistance associated protein 2 (Mrp2). BSP is transported into hepatocytes by OATPs and, after conjugation to glutathione, is excreted into bile by Mrp2. It was found to inhibit the aldo-keto reductase ARK1C20.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substrate- and pH-specific antifolate transport mediated by organic anion-transporting polypeptide 2B1 (OATP2B1-SLCO2B1).
Visentin M
Molecular Pharmaceutics, 81(2), 134-142 (2012)
Jing-Dong Zhou et al.
Journal of cellular and molecular medicine, 21(8), 1468-1481 (2017-04-30)
Promoter hypermethylation-mediated inactivation of ID4 plays a crucial role in the development of solid tumours. This study aimed to investigate ID4 methylation and its clinical relevance in myeloid malignancies. ID4 hypermethylation was associated with higher IPSS scores, but was not
Permeation of Boswellia extract in the Caco-2 model and possible interactions of its constituents KBA and AKBA with OATP1B3 and MRP2
Philip Kruger
European Journal of Pharmaceutical Sciences, 36 (2009)
Organic Anion Transporting Polypeptide Mediates Organic Anion/HCO3- Exchange*
Lisa M
The Journal of Biological Chemistry (1997)
Sreenivasulu Dasari et al.
Ecotoxicology and environmental safety, 182, 109416-109416 (2019-07-14)
The main objectives of this study were to purify the glutathione S-transfereses (GSTs) and assess the effect of high doses of acrylamide (ACR) on male albino Wistar rat liver, kidney, testis and bran GST activities, and expression analysis of GST.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service