Skip to Content
MilliporeSigma
Sulfobromophthalein disodium salt hydrate used to study hepatocyte transport functions

Sulfobromophthalein disodium salt hydrate

Sulfobromophthalein disodium salt hydrate used to study hepatocyte transport functions
Synonym(s):
BSP, Bromsulfalein sodium disodium salt hydrate
Empirical Formula (Hill Notation):
C20H8Br4Na2O10S2 · xH2O
CAS Number:
Molecular Weight:
838.00 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1