Skip to Content
MilliporeSigma

M-025

Methamphetamine/Cocaine/Heroin solution

250 μg/mL each component in acetonitrile, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Cocaine , Heroin , (±)-Methamphetamine

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), estupefaciente/psicótropo (Spain); Decreto Lei 15/93: Tabela IA/IIB (Portugal), kontrollierte Droge in Deutschland

concentration

250 μg/mL each component in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

room temp

General description

A certified Snap-N-Spike® reference solution of routinely tested drugs methamphetamine, cocaine, and heroin. Addicts often administer these drugs together as a way to increase the intensity of a “high”. This analytical reference standard is applicable in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Analyte

Description

  • (±)-Deoxyephedrine hydrochloride

  • Heroin

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

35.6 °F - closed cup

Flash Point (°C)

2 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library