Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR2684

Supelco

Cyproheptadine Hydrochloride

pharmaceutical secondary standard, certified reference material

Synonym(s):

Cyproheptadine hydrochloride sesquihydrate, 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-piperidine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N · HCl · 1.5H2O
CAS Number:
Molecular Weight:
350.88
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C3280000
traceable to USP 1161000

API family

cyproheptadine

form

solid

packaging

pkg of 500 mg
pkg of

mp

165 °C (dec.) (lit.)

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

O.O.O.Cl.CN1CCC(\CC1)=C2\c3ccccc3C=Cc4ccccc24.CN5CCC(\CC5)=C6\c7ccccc7C=Cc8ccccc68

InChI

1S/2C21H21N.ClH.3H2O/c2*1-22-14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21;;;;/h2*2-11H,12-15H2,1H3;1H;3*1H2

InChI key

PXCSXCDWTUPVIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service