Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1054

Supelco

Riboflavin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(−)-Riboflavin, Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
34058005
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. R060000
traceable to USP 1603006

API family

riboflavin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

290 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

Riboflavin may also be used as a reference standard for the determination of the analyte in cooked sausages and energy drinks by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0268 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid method for determination of riboflavin in urine by high-performance liquid chromatography
Smith DM, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 182(3), 285-291 (1980)
Determination of total riboflavin in cooked sausages
Valls F, et al.
Journal of Agricultural and Food Chemistry, 47(3), 1067-1070 (1999)
Simultaneous determination of riboflavin, pyridoxine, nicotinamide, caffeine and taurine in energy drinks by planar chromatography-multiple detection with confirmation by electrospray ionization mass spectrometry
Aranda M and Morlock G
Journal of Chromatography A, 1131(1-2), 253-260 (2006)
Zisis Gatzioufas et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(12), 846-848 (2013-10-31)
To investigate the effect of high-fluence corneal collagen cross-linking (CXL) with riboflavin and ultraviolet-A in the management of progressive keratoconus. Preliminary results from a prospective cohort study. Seven eyes from 7 patients with progressive keratoconus subjected to CXL were included.
Carol P Wilson et al.
Hypertension (Dallas, Tex. : 1979), 61(6), 1302-1308 (2013-04-24)
Intervention with riboflavin was recently shown to produce genotype-specific lowering of blood pressure (BP) in patients with premature cardiovascular disease homozygous for the 677C→T polymorphism (TT genotype) in the gene encoding the enzyme methylenetetrahydrofolate reductase (MTHFR). Whether this effect is

Articles

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service