Skip to Content
MilliporeSigma
Cyproheptadine Hydrochloride pharmaceutical secondary standard, certified reference material

Cyproheptadine hydrochloride

Cyproheptadine Hydrochloride pharmaceutical secondary standard, certified reference material
Synonym(s):
Cyproheptadine hydrochloride sesquihydrate, 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-piperidine hydrochloride hydrate
Empirical Formula (Hill Notation):
C21H21N · HCl · 1.5H2O
CAS Number:
Molecular Weight:
350.88

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade