Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1790

Supelco

Eicosapentaenoic Acid Ethyl Ester

Pharmaceutical Secondary Standard: Certified Reference Material

Synonym(s):

cis-5,8,11,14,17-Eicosapentaenoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
UNSPSC Code:
12352211
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. E0085000
traceable to USP 1234307

API family

eicosapentaenoic acid

form

liquid

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

O(CC)C(=O)CCC\C=C\C\C=C\C\C=C\C\C=C\C\C=C\CC

InChI

1S/C22H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-4-2/h5-6,8-9,11-12,14-15,17-18H,3-4,7,10,13,16,19-21H2,1-2H3/b6-5+,9-8+,12-11+,15-14+,18-17+

InChI key

SSQPWTVBQMWLSZ-LRKAYDMASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Docosahexaenoic Acid Ethyl Ester is the ethyl ester of docosahexaenoic acid, an essential omega-3 fatty acid.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Development and validation of a reverse phase high-performance liquid chromatography-tandem mass spectrometry (RP-HPLC-MS/MS) method to determine eicosapentaenoic acid and docosahexaenoic acid in their ethyl ester forms
  • Simulated moving bed (SMB) chromatographic separation of ethyl esters of eicosapentaenoic acid and docosahexaenoic acids

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2773 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>320.0 °F

Flash Point(C)

> 160 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service