Skip to Content
MilliporeSigma

MAK134

Ketone Body Assay

sufficient for 200 tests (UV)

Synonym(s):

Ketone Quantification Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 200 tests (UV)

feature

This kit is sufficient for up to 100 assays of 3-hydroxybutyric acid (BOH) and up to 100 assays of acetoacetic acid (AcAc) in 96 well plates.

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; neonatal diseases; pediatric diseases; cancer

storage temp.

−20°C

General description

Ketone bodies, 3-hydroxybutyric acid (BOH) and acetoacetic acid (AcAc), are produced in the liver primarily from oxidation of fatty acids, and are normally present at low concentrations in urine and blood. Increased ketone concentrations in the blood may lead to metabolic acidosis, which has been associated with diabetes, childhood hypoglycemia, growth hormone deficiency, alcohol or salicylate intoxication, and inborn errors of metabolism.

Application

Ketone Body Assay has been used to measure the release of ketone bodies in the human liver cancer cell line HepG2 culture medium.
Suitable for the detection of ketone bodies (acetoacetic acid and 3-hydroxybutyric acid) in serum, plasma, urine, and other biological samples.

Biochem/physiol Actions

In this kit, AcAc and BOH levels are determined using an enzymatic assay based on 3-hydroxybutyrate dehydrogenase catalyzed reactions, in which the change in NADH absorbance measured at 340 nm is directly related to either the AcAc and BOH concentrations.

Features and Benefits

Compatible with high-throughput handling systems


Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library