Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1575

Supelco

Propantheline Bromide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Propantheline bromide, (2-Hydroxyethyl)diisopropylmethylammonium bromide xanthene-9-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C23H30NO3Br
CAS Number:
Molecular Weight:
448.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1570508

API family

propantheline

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].CC(C)[N+](C)(CCOC(=O)C1c2ccccc2Oc3ccccc13)C(C)C

InChI

1S/C23H30NO3.BrH/c1-16(2)24(5,17(3)4)14-15-26-23(25)22-18-10-6-8-12-20(18)27-21-13-9-7-11-19(21)22;/h6-13,16-17,22H,14-15H2,1-5H3;1H/q+1;/p-1

InChI key

XLBIBBZXLMYSFF-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Propantheline Bromide belongs to the class of quaternary ammonium compounds and is a synthetic anticholinergic drug. It is widely used as an oral agent and it exhibits antimuscarinic effect in the lower urinary tract.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Inhibits action of acetylcholine at post-ganglionic synapses; muscarinic acetylcholine receptor antagonist; antispasmodic; reduces gastric acid secretion.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4491 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cohen's Pathways of the Pulp Expert Consult - E-Book (2010)
Female Urology E-Book: Text with DVD (2008)
Andrés Montoya et al.
Frontiers in cellular and infection microbiology, 8, 350-350 (2018-10-20)
Introduction: In cutaneous leishmaniasis, the host immune response is responsible for the development of skin injuries but also for resolution of the disease especially after antileishmanial therapy. The immune factors that participate in the regulation of inflammation, remodeling of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service