Skip to Content
MilliporeSigma
All Photos(1)

Documents

B7522

Supelco

Benfluorex hydrochloride

analytical standard

Synonym(s):

N-(1-Methyl-2-(3-[trifluoromethyl]-phenyl)ethyl]aminoethanol benzoate ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F3NO2 · HCl
CAS Number:
Molecular Weight:
387.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(Cc1cccc(c1)C(F)(F)F)NCCOC(=O)c2ccccc2

InChI

1S/C19H20F3NO2.ClH/c1-14(12-15-6-5-9-17(13-15)19(20,21)22)23-10-11-25-18(24)16-7-3-2-4-8-16;/h2-9,13-14,23H,10-12H2,1H3;1H

InChI key

NLOALSPYZIIXEO-UHFFFAOYSA-N

Gene Information

human ... SOAT1(6646)

General description

Benfluorex hydrochloride salt is a hypolipidaemic compound with lipid and glucose lowering effects.
It is used in the treatment of impaired carbohydrate metabolism, atherogenic metabolic disorders and in the suppression of appetite.

Application

Benfluorex hydrochloride may be used as a reference standard for characterization of human urinary metabolites of benfluorex by gas chromatography–electron ionization–mass spectrometry (GC-EI-MS) as well as liquid chromatography–electrospray ionization–high resolution/high accuracy tandem mass spectrometry (LC-ESI-MS/MS) method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Agnès Fournier et al.
Pharmacoepidemiology and drug safety, 21(4), 343-351 (2012-02-10)
To estimate the number of deaths from valvular insufficiency caused by the use of the fenfluramine-derivative benfluorex during the period 1976-2009 in France. Our calculation was based on (i) the exposure level to benfluorex in the French population, derived from
Thermal and X-ray powder diffraction structural characterization of two benfluorex hydrochloride polymorphs.
Maccaroni E, et al.
Journal of Pharmaceutical and Biomedical Analysis, 53(1), 1-6 (2010)
Screening for benfluorex and its major urinary metabolites in routine doping controls.
Thevis M, et al.
Analytical and Bioanalytical Chemistry, 401(2), 543-551 (2011)
Nam Sook Kim et al.
Rapid communications in mass spectrometry : RCM, 35(3), e8989-e8989 (2020-10-27)
Recently, illegal dietary supplements containing unauthorized compounds have been seized and internationally publicized with a warning to avoid their consumption. This adulteration can be a serious threat to public health because of insufficient and reliable safety data as well as
Mediator scandal engulfs French compensation body.
Asher Mullard
Lancet (London, England), 381(9880), 1803-1803 (2013-05-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service