Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1360

Supelco

Potassium Iodide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Potassium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1548280

vapor pressure

1 mmHg ( 745 °C)

API family

potassium iodide

CofA

current certificate can be downloaded

packaging

pkg of 5 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

681 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium Iodide is well-known for being the first drug in the successful treatment of human sporotrichosis. Today it is widely used for the treatment of various kinds of skin inflammation disorders such as erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet′s syndrome. On account of its low cost, it continues to be used for cutaneous and lymphocutaneous diseases. It is also a part of the electrolyte of dye sensitized solar cells (DSSC).
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1464 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium iodide capsule treatment of feline sporotrichosis.
Reis EG, et al.
Journal of Feline Medicine and Surgery, 14(6), 399-404 (2012)
Effects of potassium iodide (KI) on crystallinity, thermal stability, and electrical properties of polymer blend electrolytes (PVC/PEO: KI)
Nadimicherla R, et al.
Solid State Ionics, 278(6), 260-267 (2015)
Sungwook Choung et al.
Environmental science & technology, 48(16), 9684-9691 (2014-08-05)
Bentonite, which is used as an engineered barrier in geological repositories, is ineffective for sorbing anionic radionuclides because of its negatively charged surface. This study modified raw bentonite using a cationic surfactant (i.e., hexadecyltrimethylammonium [HDTMA]-Br) to improve its sorption capability
J B Sterling et al.
Journal of the American Academy of Dermatology, 43(4), 691-697 (2000-09-27)
Potassium iodide (KI) is a useful drug in the dermatologic armamentarium. It is successfully used for inflammatory dermatoses, most notably erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet's syndrome. KI is also successfully used for cutaneous
Natchanon Amornthammarong et al.
The Analyst, 139(20), 5263-5270 (2014-08-20)
A new system requiring greatly reduced operator intervention has been developed for the determination of dissolved inorganic carbon concentration in marine waters. Based on a coulometric method, the system has an accuracy and precision comparable to more complex and expensive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service