Skip to Content
MilliporeSigma

1548280

USP

Potassium iodide

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor pressure

1 mmHg ( 745 °C)

API family

potassium iodide

manufacturer/tradename

USP

mp

681 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Potassium iodide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Potassium iodide


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Oral

target_organs

Thyroid

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library