MilliporeSigma
  • A simplified coulometric method for multi-sample measurements of total dissolved inorganic carbon concentration in marine waters.
A simplified coulometric method for multi-sample measurements of total dissolved inorganic carbon concentration in marine waters.
The Analyst (2014-08-20)
Natchanon Amornthammarong, Peter B Ortner, James Hendee, Ryan Woosley
ABSTRACT
A new system requiring greatly reduced operator intervention has been developed for the determination of dissolved inorganic carbon concentration in marine waters. Based on a coulometric method, the system has an accuracy and precision comparable to more complex and expensive methods currently employed. A syringe pump equipped with a 12-port distribution valve is used to precisely dispense an acid solution and sample into a gas stripper. The system can autonomously measure eight discrete samples in duplicate or triplicate with no operator input. The best precision (%RSD) obtained was 0.022% (n = 14) or less than ±1.0 μmol kg(-1). The system is calibrated against a certified reference material (CRM). Average offset from the CRM was 1.2 μmol kg(-1). Sample throughput was 4 samples per h. Carryover effects are negligible but field sample analyses suggest that prefiltering may be necessary in highly turbid waters.
Sigma-Aldrich
Potassium iodide, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium iodide, SAJ first grade, ≥99.5%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%