Skip to Content
MilliporeSigma
All Photos(1)

Documents

NISTRM8551

USGS26 (nitrogen isotopes in ammonium sulfate)

NIST® RM 8551

Synonym(s):

Ammonium sulfate, Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

isotopic purity

 15N

manufacturer/tradename

NIST®

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Reference/Informational value is provided for the following analytes.
Nitrogen (15N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service