Skip to Content
MilliporeSigma
All Photos(1)

Documents

NISTRM8539

NBS 22 oil (carbon and hydrogen isotopes in oil)

NIST RM 8539

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

isotopic purity

 2H
 13C

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

Reference/Informational value is provided for the following analytes.Carbon (13C), Hydrogen (2H)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 - 651.2 °F

Flash Point(C)

98 - 344 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service