Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST614

Trace elements in glass

NIST® SRM® 614

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Quality Level

packaging

pkg of 4 wafers

manufacturer/tradename

NIST®

application(s)

glass & ceramic

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A unit of SRM 614 comprises four wafers, each sliced to a thickness of 3 mm from a hand-pulled rod. The wafers have an ovalto circular cross-section, with a nominal diameter ranging from 12 mm to 14 mm.

SRM 614_cert

SRM 614_SDS

Application

The SRM is intended for use in the chemicalmethods employed to analyze trace elements in glass.

Features and Benefits

  • Accompanied with certified mass fraction values for each elements in trace elements in glass.
  • The reference and information values of elements are reported as mass fraction.
  • The NIST certificate is provided with expiration certificate, storage, safety, handling,use, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Copper (Cu), Lead (Pb), Potassium (K), Rubidium (Rb), Silver (Ag), Strontium (Sr), Thorium (Th), Uranium (U)

Reference/Informational value is provided for the following analytes.
Boron (B), Cobalt (Co), Europium (Eu), Iron (Fe), Lanthanum (La), Scandium (Sc), Thallium (Th), Titanium (Ti), Strontium (87Sr/86Sr), Antimony (Sb), Cadmium (Cd), Gallium (Ga), Gold (Au), Nickel (Ni), Uranium (235U/UTotal)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service