Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST3112A

Chromium (Cr) standard solution

NIST® SRM® 3112a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

A unit of the SRM contains five 10 mL sealed borosilicate glass ampoules, consisting of an acidified aqueous solution. The solution is prepared gravimetrically to contain a known mass fraction of chromium and includes nitric acid at a mass fraction of approximately 15%. For more information, please refer to the SDS and COA.

SRM 3112A_cert

SRM 3112A_SDS

Application

Chromiumstandard solution is intended for use as aprimary calibration standard for the quantitative determination of chromium.

Features and Benefits

  • NIST Certified Mass fraction value of Chromium is basedon gravimetric preparation using high-purity chromium metal and inductivelycoupled plasma optical emission spectrometry (ICP-OES).
  • Expiration details, instructions for use, handling, andstorage are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Chromium

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service