Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3144

Rhodium (Rh) standard solution

NIST® SRM® 3144

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

One unit of SRM 3144 contains five sealed borosilicate glass ampoules, each containing 10 mL of an acidified aqueous solution prepared from high-purity ammonium hexachlororhodate (III). This solution contains approximately 6% hydrochloric acid by mass fraction.

SRM 3144_cert

SRM 3144_SDS

Application

Rhodium (Rh) standard solution SRM is intended for use as a calibration standard for quantitative determination of rhodium.

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values of Rhodium, which is obtained through inductively coupled plasma optical emission spectrometry (ICP-OES).
  • Certified value is metrologically traceable to the derived SI unit for mass fraction, expressed as milligrams per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Rhodium (Rh)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service