Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST979

Chromium isotopic standard

NIST® SRM® 979

Synonym(s):

Chromium(III) nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
400.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

isotopic purity

 50Cr
 52Cr
 53Cr
 54Cr

packaging

pkg of 0.25 g

manufacturer/tradename

NIST®

mp

60 °C (lit.)

format

matrix material

SMILES string

[Cr+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cr.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

GVHCUJZTWMCYJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Features and Benefits

Measurements of hydrated chromium nitrate areconducted using single filament (platinum) mass spectrometry.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Chromium-50, Chromium-52, Chromium-53, Chromium-54

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service