Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3101A

Aluminum (Al) standard solution

NIST® SRM® 3101a

Synonym(s):

Aluminum Calibration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of aluminum. A unit of SRM 3101a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of aluminum. The solution contains nitric acid at a volume fraction of approximately 10 %, equivalent to an amount of substance concentration (molarity) of approximately 1.6 mol/L. For more information, please refer to the SDS and COA.

SRM 3101A_cert

SRM 3101A_SDS

Application


  • Aluminum standard solution: Aluminum standard solutions are essential in trace metal analysis, particularly in pharmaceutical quality control where accuracy and precision are paramount. These standards are used to calibrate instruments such as ICP-MS, ensuring reliable detection and quantification of trace metals in pharmaceutical products. The use of aluminum ICP standard solutions is crucial for maintaining compliance with regulatory standards and ensuring the safety and efficacy of pharmaceuticals. They are also utilized in biochemical research as high-purity reference materials to study metal-related biochemical processes (Jafari et al., 2024; Barsola et al., 2024).

Other Notes

Certified for the analytes listed below.
Aluminium (Al)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service