Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST194A

Ammonium dihydrogen phosphate

NIST® SRM® 194a

Synonym(s):

Ammonium phosphate monobasic, mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 90 g

manufacturer/tradename

NIST®

mp

190 °C (dec.) (lit.)

application(s)

agriculture

format

neat

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a highly purified and homogeneous lot of crystalline ammonium dihydrogen phosphate (NH4H2PO4). It is intended primarily for use as a working standard in the calibration and standardization of procedures employed in the fertilizer industry for the determination of ammoniacal nitrogen and phosphorus. For more information, please refer to the SDS and COA.

SRM 194A_cert

SRM 194A_SDS

Application

  • Improving fourth harmonic generation performance by elevating the operation temperature of ADP crystal: This study highlights the enhancement of fourth harmonic generation in ADP crystals by manipulating their operational temperature, demonstrating a crucial application in optical technologies (Liu et al., 2024).
  • Growth and Temperature Properties of the Phase-Matching Angle of 23% Deuterated ADP Crystal: This research explores the phase-matching properties of deuterated ADP crystals, critical for improving the efficiency of nonlinear optical devices (Sun et al., 2024).
  • Comparison of hydrogen vacancies in KDP and ADP crystals: This paper uses a combination of density functional theory and experimental approaches to study hydrogen vacancies in ADP crystals, providing insights into the structural properties that influence their optical performance (Sui et al., 2019).
  • Structural, spectroscopic, optical, dielectric and mechanical study of pure and l-Proline doped ammonium dihydrogen phosphate single crystals: This comprehensive study addresses how doping ADP crystals with l-proline alters their physical properties, suggesting applications in both optical fields and material science (Hasmuddin et al., 2014).

Other Notes

Certified for the analytes listed below.
Nitrogen (N), Phosphorus (P)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Fluorine (F), Neodymium (Nd), Sulfur (S), Antimony (Sb), Gadolinium (Gd), Nickel (Ni), Tantalum (Ta), Arsenic (As), Gallium (Ga), Niobium (Nb), Tellurium (Te), Barium (Ba), Germanium (Ge), Osmium (Os), Terbium (Tb), Beryllium (Be), Gold (Au), Palladium (Pd), Thallium (Th), Bismuth (Bi), Hafnium (Hf), Latinum (La), Thorium (Th), Boron (B), Holmium (Ho), Potassium (K), Thulium (Tm),Bromine (Br), Indium (In), Praseodymium (Pr), Tin (Sn), Cadmium (Cd), Iodine (I), Rhenium (Re), Titanium (Ti), Calcium (Ca), Iridium (Ir) Rhodium (Rh), Tungsten (W), Cerium (Ce), Iron (Fe) Rubidium (Rb), Uranium (U), Cesium (Ce), Lanthanum (La), Ruthenium (Ru), Vanadium (V), Chlorine (Cl), Lead (Pb), Samarium (Sm), Ytterbium (Yb), Chromium (Cr), Lithium (Li), Scandium (Sc), Yttrium (Yt), Cobalt (Co), Lutetium (Lu), Selenium (Se), Zinc (Zn), Copper (Cu), Magnesium (Mg), Silicon (Si), Zirconium (Zr), Dysprosium (Dy), Manganese (Mn), Silver (Ag), Erbium (Er) Mercury (Hg), Sodium (Na), Europium (Eu), Molybdenum (Mo), Strontium (Sr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service