Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01126

Supelco

Ammonium dihydrogen phosphate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ammonium dihydrogen phosphate, Ammonium biphosphate, Ammonium phosphate monobasic, Primary ammonium phosphate, Monoammonium orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)H2PO4
CAS Number:
Molecular Weight:
115.03
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-764-5
NACRES:
NA.21
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

0.066 hPa ( 125 °C)

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

potency

5750 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

impurities

≤0.005% Insoluble matter

pH

3.8-4.4 (20 °C, 50 g/L in H2O)

mp

190 °C

density

1.80 g/cm3 at 20 °C

bulk density

800‑1100 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
nitrate (NO3-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.001%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[P](=O)([O-])([O-])[O-].[N+H4].[H+].[H+]

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis and characterization of hydroxyapatite nanoparticles from calcium hydroxide fouled with gases evolved from smokestack of glass industry.: This research explores the synthesis of hydroxyapatite nanoparticles using ammonium dihydrogen phosphate, detailing the process and analytical chemistry techniques involved. The study provides insights into the potential environmental and industrial applications of this method, offering an innovative approach to materials synthesis (Abdelmoaty and Mousa, 2024).

  • Acid etching-induced nanocutting of LaNiO(3) transplanting self-assembled photodiode array-like LaNiO(3)/N,P-RGO nanoreactor for efficient photocatalytic hydrogen evolution.: This article discusses the use of ammonium dihydrogen phosphate in creating LaNiO(3) based nanoreactors through acid etching techniques. The resulting materials show promising applications in photocatalysis and hydrogen production, illustrating the compound′s utility in renewable energy technologies (Zou et al., 2024).

  • Sustainable and Environmentally Friendly Microwave Synthesis of Nano-Hydroxyapatite from Decarbonized Eggshells.: This study utilizes ammonium dihydrogen phosphate in a green chemistry process to synthesize nano-hydroxyapatite from eggshells, emphasizing the environmental benefits and sustainability of this approach in materials science (Mahmoud, 2024).

  • A Self-Powered Biochemical Sensor for Intelligent Agriculture Enabled by Signal Enhanced Triboelectric Nanogenerator.: This innovative research uses ammonium dihydrogen phosphate in the development of a self-powered biochemical sensor for agricultural applications, highlighting its role in enhancing device sensitivity and sustainability (Gao et al., 2024).

  • Therapeutic drug monitoring of vancomycin in surgical patients through a validated HPLC method.: In this clinical study, ammonium dihydrogen phosphate is employed as part of a high-performance liquid chromatography (HPLC) method for monitoring vancomycin in surgical patients, demonstrating its crucial role in improving analytical accuracy and patient care (Akram et al., 2024).

Linkage

Replaces: AX1350; AX1350-1; AX1350-20

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water, 25 °C): 3.8 - 4.4
pH-value (2.3%; water): about 4.2
Chloride (Cl): ≤ 0.0005 %
Nitrate (NO₃): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service