Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1921B

IR transmission wavelength (polystyrene film)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 1 card

manufacturer/tradename

NIST®

application(s)

general analytical

General description

A unit of SRM 1921b consists of a matte finish polystyrene film approximately 38 µm thick, with a 25 mm diameter of exposed area, centered 38 mm from the bottom of a cardboard holder, which is 5 cm × 11 cm × 0.2 cm in size.

The certification of SRM 1921b is valid, within the measurement uncertainty specified, provided the SRM is handled and stored in accordance with the instructions given in the accompanying certificate (see “Instructions for Storage, Handling, and Use”).

SRM 1921B_cert

SRM 1921B_SDS

Application

This Standard Reference Material (SRM) is intended primarily for use in calibrating the wavelength/wavenumber scale of spectrophotometers in the infrared (IR) spectral region from 3.2 µm to 18.5 µm (540 cm-1 to 3125 cm-1).

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service