Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST1479

Polystyrene (narrow molecular weight distribution)

NIST® SRM® 1479

Synonym(s):

Polystyrene, PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

beads

packaging

pkg of 2 g

manufacturer/tradename

NIST®

application(s)

chemicals and industrial polymers

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for the calibration and evaluation of instruments used in polymer technology and science for the determination of the molecular weight of polystyrene. The SRM is supplied as white pellets of polystyrene in a 2 gram unit. For more information, please refer to the SDS and COA.

SRM 1479_cert

SRM 1479_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Polystyrene

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service