Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1019B

Glass (particle size)

NIST® SRM® 1019b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

beads

packaging

pkg of 200 g

manufacturer/tradename

NIST®

application(s)

glass & ceramic

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating and calibrating particle size measurement instrumentation covering 750 μm to 450 μm range. Typical use is in the evaluation of wire-cloth test sieives in the range from No. 20 (850 μm) through No. 10 (2000 μm). The SRM consist of a single bottle containing approximately 200 g of sold spherical soda-lime glass beads. For more information, please refer to the SDS and COA.

SRM 1019B_cert

SRM 1019B_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service