Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1985

Thermal Spray Powder - Particle Size Distribution Tungsten Carbide/Cobalt (Spheroidal)

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

powder

packaging

pkg of 14 g

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable
electron microscopy: suitable

storage temp.

15-25°C

General description

This Standard Reference Material (SRM) is intended primarily for use in the calibration of equipment used to measure particle size distributions (PSD) in the 18μm to 55μm range. SRM 1985 consists of a single bottle containing approximately 14 g of tungsten carbide/cobalt powder.

SRM 1985_cert

SRM 1985_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service