Skip to Content
MilliporeSigma
All Photos(2)

Documents

NIST2066

K-411 Glass microspheres

NIST® SRM® 2066

Synonym(s):

Glass spheres

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

powder

packaging

pkg of 50 mg

manufacturer/tradename

NIST®

density

1.1 g/mL at 25 °C (lit.)

application(s)

glass & ceramic

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended for use as a standard for the quantitative microanalysis of particles and for the development of particle matrix correction procedures. The elements present in these microspheres, common to many classes of particles, are silicon, calcium, magnesium, iron, and oxygen. SRM 2066 consists of approximately 50 mg of glass microspheres of known composition and size (1 μm to 40 μm) with minimal microheterogeneity. For more information, please refer to the COA and SDS.

SRM 2066_cert

SRM 2066_SDS

Other Notes

Certified for the analytes listed below.
Silicon (Si), Calcium (Ca), Magnesium (Mg), Iron (Fe), Oxygen (O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service