Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2250000

Dinoprostone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Prostaglandin E2, (5Z,11α,13E,15S)-11,15-Dihydroxy-9-oxoprosta-5,13-dienoic acid, Dinoprostone, PGE2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
Beilstein:
4709356
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dinoprostone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O[C@@H]1CC([C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/[C@@H](O)CCCCC)=O

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,19,21,23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,19+/m0/s1

InChI key

XEYBRNLFEZDVAW-ARSRFYASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dinoprostone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Most biologically active prostaglandin.
Most biologically active prostaglandin. PGE2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation; regulates adenylyl cyclase. Tumor cells that over-express cyclooxygenase 2 display increased invasiveness, angiogenesis, and resistance to apoptosis that may be due to the PGE2-induced expression of angiogenic factors and stabilization of the anti-apoptotic protein, survivin.The effect of PGE2 on the immune system is mixed. It inhibits T cell activation in vitro, suggesting it is an immunosuppressant. However, in vivo, it appears to effect expansion of the Th17 subset and differentiation of the Th1 subset of T helper cells, marking it as an immunoactivator.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiki Kanazawa et al.
PloS one, 9(8), e104676-e104676 (2014-08-08)
Pressure ulcers are characterized by chronicity, which results in delayed wound healing due to pressure. Early intervention for preventing delayed healing due to pressure requires a prediction method. However, no study has reported the prediction of delayed healing due to
Sonja I Gringhuis et al.
Nature communications, 5, 5074-5074 (2014-10-04)
Dendritic cells (DCs) orchestrate antibody-mediated responses to combat extracellular pathogens including parasites by initiating T helper cell differentiation. Here we demonstrate that carbohydrate-specific signalling by DC-SIGN drives follicular T helper cell (TFH) differentiation via IL-27 expression. Fucose, but not mannose
Yumeng Mao et al.
Cancer research, 73(13), 3877-3887 (2013-05-02)
Tumors can suppress the host immune system by employing a variety of cellular immune modulators, such as regulatory T cells, tumor-associated macrophages, and myeloid-derived suppressor cells (MDSC). In the peripheral blood of patients with advanced stage melanoma, there is an
Takayuki Nakagawa
Hearing research, 276(1-2), 27-33 (2011-02-08)
Prostaglandins are one of the major groups of chemical mediators in the mammalian body. Among prostaglandins, prostaglandin E2 (PGE2) is the most abundant prostanoid in humans and involved in regulating many different fundamental biological functions. PGE2 signaling is mediated by
Sanjiv Dhingra et al.
Circulation, 128(11 Suppl 1), S69-S78 (2013-10-18)
Allogeneic mesenchymal stem cells (MSCs) were immunoprivileged early after cardiac implantation and improved heart function in preclinical and clinical studies. However, long-term preclinical studies demonstrated that allogeneic MSCs lost their immunoprivilege and were rejected in the injured myocardium, resulting in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service