Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000054

Alprostadil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Prostaglandin E1, (11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
Beilstein:
5294062
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

alprostadil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1

InChI key

GMVPRGQOIOIIMI-DWKJAMRDSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alprostadil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristine J Guleserian et al.
The Journal of thoracic and cardiovascular surgery, 145(1), 206-213 (2012-12-19)
Bilateral pulmonary artery banding with or without ductal stenting has been performed as a resuscitative intervention for patients considered at too high risk for conventional single ventricle palliation. The purpose of the present study was to determine the outcomes using
K Schrör et al.
VASA. Zeitschrift fur Gefasskrankheiten, 33(3), 119-124 (2004-10-06)
The mechanisms of anti-ischemic effects of PGE1 in patients with peripheral arterial occlusive disease (PAD) are probably complex and clearly not limited to a direct vasodilator action. In addition to the known effects on blood flow, viscosity, fibrinolysis and platelet
Pierre Costa et al.
Drugs, 72(17), 2243-2254 (2012-11-23)
The global burden of erectile dysfunction (ED) is increasing. It is estimated that 8-19% of men in Europe have ED and that by 2025 the prevalence of ED worldwide will reach 322 million. The gold standard therapy for ED is
Takashi Onoe et al.
Transplantation, 95(12), 1521-1527 (2013-04-20)
Small-for-size syndrome remains the greatest limiting factor of expanding segmental liver transplantation from living donors. Portal hyperperfusion is considered to substantially contribute to small-for-size syndrome. We investigated the impact of continuous portal infusion of prostaglandin E1 (PGE1) on small-for-size grafts
Pot economy in the synthesis of prostaglandin A1 and E1 methyl esters.
Yujiro Hayashi et al.
Angewandte Chemie (International ed. in English), 52(12), 3450-3452 (2013-02-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service