Skip to Content
MilliporeSigma

1213001

USP

Dinoprost tromethamine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Prostaglandin F tris salt, (5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt, PGF−Tris

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H34O5 · C4H11NO3
CAS Number:
Molecular Weight:
475.62
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4087514
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dinoprost

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(CO)(CO)CO.[H][C@]1(C\C=C/CCCC(O)=O)[C@@H](O)C[C@@H](O)[C@]1([H])\C=C\[C@@H](O)CCCCC

InChI

1S/C20H34O5.C4H11NO3/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25;5-4(1-6,2-7)3-8/h4,7,12-13,15-19,21-23H,2-3,5-6,8-11,14H2,1H3,(H,24,25);6-8H,1-3,5H2/b7-4-,13-12+;/t15-,16+,17+,18-,19+;/m0./s1

InChI key

IYGXEHDCSOYNKY-RZHHZEQLSA-N

Gene Information

human ... PTGFR(5737)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dinoprost tromethamine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dinoprost Tromethamine Injection

Biochem/physiol Actions

Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds.
Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. A positive feedback loop between endometrial PG F and luteal oxytocin is responsible for completion of luteolysis.
Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. Also stimulates contraction of smooth muscle, such as in bronchi, with possible involvement in asthma. This effect is at least partially mediated through tachykinin release.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library