Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1500000

Chlordiazepoxide hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-benzodiazepine 4-oxide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClN3O · HCl
CAS Number:
Molecular Weight:
336.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlordiazepoxide

manufacturer/tradename

EDQM

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CNC1=Nc2ccc(Cl)cc2C(c3ccccc3)=[N+]([O-])C1

InChI

1S/C16H14ClN3O.ClH/c1-18-15-10-20(21)16(11-5-3-2-4-6-11)13-9-12(17)7-8-14(13)19-15;/h2-9H,10H2,1H3,(H,18,19);1H

InChI key

DMLFJMQTNDSRFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlordiazepoxide hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholestatic jaundice associated with chlordiazepoxide hydrochloride (Librium) therapy. Report of a case and review of the literature.
K J Lo et al.
The American journal of digestive diseases, 12(8), 845-849 (1967-08-01)
B C Ginsburg et al.
British journal of pharmacology, 171(14), 3499-3510 (2014-04-05)
Drugs that more potently or effectively reduce ethanol-maintained behaviour versus an alternative are considered selective and are considered promising pharmacotherapies for alcoholism. Such results are often obtained using separate groups or multiple schedules where ethanol and the alternative are available
Lidia Manzo et al.
Behavioural brain research, 278, 90-97 (2014-09-23)
Rats increased preference for ethanol after sessions of appetitive extinction, but not after acquisition (reinforced) sessions (Manzo et al., 2014). Drinking was not influenced by appetitive extinction in control groups with postsession access to water, rather than ethanol. Because ethanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service