Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR705

Linde Type A Zeolite

BCR®, certified reference material

Synonym(s):

Zeolite, Linde Type A

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

Application

Linde type A zeolite has been used as a host in the preparation of drug delivery system containing 5-fluorouracil.

Analysis Note

For more information please see:
BCR705

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of different silica microporous structures as drug delivery systems for in vitro models of solid tumors
Vilaca N, et al.
Royal Society of Chemistry Advances, 7, 13104-13111 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service