Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382280

Sigma-Aldrich

Silver-exchanged zeolite

granular, +20 mesh

Synonym(s):

Ag

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag84Na2[(AlO2)86(SiO2)106] · xH2O
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

granular

Quality Level

composition

Ag, 35%

reaction suitability

core: silver
reagent type: catalyst

particle size

+20 mesh

mp

>500 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Silver-exchanged zeolite selectively removes halogens from gas streams and also serves as catalyst.

Application

Silver-exchanged zeolite is used:
  • As a catalyst in the low-temperature CO isotopic scrambling reaction.
  • To capture radioiodine from the vapor phase, which is then thermally converted into silver sodalite.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver iodide sodalite--Wasteform/Hip canister interactions and aqueous durability
Maddrell ER, et al.
Journal of Nuclear Materials, 517, 71-79 (2019)
Silver-catalyzed low-temperature CO isotopic scrambling reaction: 12 C 16 O+ 13 C 18 O? 12 C 18 O+ 13 C 16 O
Lagunov O, et al.
Physical Chemistry Chemical Physics, 14(7), 2178-2218 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service