Skip to Content
MilliporeSigma

Skip To

1.05703

Molecular sieve 1.0 nm

beads ~ 2 mm

Synonym(s):

Molecular sieve 1.0 nm, Sodium aluminium silicate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
215-283-8
NACRES:
NB.21
eCl@ss:
32049101
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

beads

surface area

800 m2/g

loss

≤5% loss on ignition, 950°C

matrix

Zeolite base material

particle size

~2 mm

pore size

1.0 nm

pH

8-11 ( in H2O)

mp

>1600 °C

bulk density

700‑750 kg/m3

storage temp.

no temp limit

SMILES string

[Si](=O)=O.[Al](=O)O[Al]=O

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Loss on ignition (950 °C): ≤ 5 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 24 %


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters