Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

95295

Supelco

Hydrogen ionophore II

Selectophore

Synonym(s):

4-Nonadecylpyridine, ETH 1907, Proton ionophore II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H43N
CAS Number:
Molecular Weight:
345.60
Beilstein:
217205
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

SMILES string

CCCCCCCCCCCCCCCCCCCc1ccncc1

InChI

1S/C24H43N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-24-20-22-25-23-21-24/h20-23H,2-19H2,1H3

InChI key

UMOIOBOWJASTRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

H+-ionophore showing a linear response from pH 2 to 9 in an optimized membrane composition

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Chao et al.
Pflugers Archiv : European journal of physiology, 411(2), 216-219 (1988-02-01)
A series of new neutral hydrogen ion carriers suitable for application in H+-selective microelectrodes is presented. One carrier (ETH 1907) proves to be superior to tridodecylamine currently very much in use. Microelectrodes based on ETH 1907 in an optimized membrane
Eskedar Ayele Angamo et al.
Journal of neurophysiology, 116(5), 2420-2430 (2016-08-26)
Astrocyte-derived lactate supports pathologically enhanced neuronal metabolism, but its role under physiological conditions is still a matter of debate. Here, we determined the contribution of astrocytic neuronal lactate shuttle for maintenance of ion homeostasis and energy metabolism. We tested for
Brian Roland Larsen et al.
The Journal of physiology, 597(2), 583-597 (2018-10-26)
Neuronal activity induces fluctuation in extracellular space volume, [K+ ]o and pHo , the management of which influences neuronal function The neighbour astrocytes buffer the K+ and pH and swell during the process, causing shrinkage of the extracellular space In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service