Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95291

Supelco

Hydrogen ionophore I - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

composition

Hydrogen ionophore I, 10.0 wt. % (95292)
2-Nitrophenyl octyl ether, 89.3 wt. % (73732)
Sodium tetraphenylborate, 0.7 wt. % (72018)

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashok Aspatwar et al.
FEBS open bio, 9(7), 1204-1211 (2019-04-30)
Carbonic anhydrases (CA) catalyze the reversible hydration of CO2 to protons and bicarbonate and thereby play a fundamental role in the epithelial acid/base transport mechanisms serving fluid secretion and absorption for whole-body acid/base regulation. The three carbonic anhydrase-related proteins (CARPs)
Lydie Cheval et al.
American journal of physiology. Renal physiology, 317(2), F435-F443 (2019-06-13)
We have recently reported that type A intercalated cells of the collecting duct secrete Na+ by a mechanism coupling the basolateral type 1 Na+-K+-2Cl- cotransporter with apical type 2 H+-K+-ATPase (HKA2) functioning under its Na+/K+ exchange mode. The first aim
Sina Ibne Noor et al.
eLife, 7 (2018-05-29)
Many tumor cells produce vast amounts of lactate and acid, which have to be removed from the cell to prevent intracellular lactacidosis and suffocation of metabolism. In the present study, we show that proton-driven lactate flux is enhanced by the
Shubha R Shanbhag et al.
The Journal of physiology, 595(2), 523-539 (2016-07-05)
The digestive tract of larval and adult Drosophila is an excellent analogue of the mammalian gut. Enterocytes of the posterior midgut are separated by septa, with no paracellular path, and therefore perform both immune and transport functions. Using microperfusion electrophysiology
Qi Wu et al.
Plant physiology, 183(3), 1331-1344 (2020-05-06)
Hydrogen gas (H2) has a possible signaling role in many developmental and adaptive plant responses, including mitigating the harmful effects of cadmium (Cd) uptake from soil. We used electrophysiological and molecular approaches to understand how H2 ameliorates Cd toxicity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service