Skip to Content
MilliporeSigma
All Photos(1)

Documents

95292

Supelco

Hydrogen ionophore I

Selectophore, function tested

Synonym(s):

Tridodecylamine, Proton ionophore I, Hydrogen ionophore I

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]3N
CAS Number:
Molecular Weight:
521.99
Beilstein:
1801669
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

vapor density

>1 (vs air)

product line

Selectophore

form

liquid

quality

function tested

refractive index

n20/D 1.4578 (lit.)

bp

220-228 °C/0.03 mmHg (lit.)

density

0.823 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C36H75N/c1-4-7-10-13-16-19-22-25-28-31-34-37(35-32-29-26-23-20-17-14-11-8-5-2)36-33-30-27-24-21-18-15-12-9-6-3/h4-36H2,1-3H3

InChI key

SWZDQOUHBYYPJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

374.0 °F

Flash Point(C)

190 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judith A Heiny et al.
The Journal of general physiology, 151(9), 1146-1155 (2019-07-20)
Ion movements across biological membranes, driven by electrochemical gradients or active transport mechanisms, control essential cell functions. Membrane ion movements can manifest as electrogenic currents or electroneutral fluxes, and either process can alter the extracellular and/or intracellular concentration of the
G T McEwan et al.
Journal of medical engineering & technology, 14(1), 16-20 (1990-01-01)
Incorporation of a ligand into polyvinylchloride allows the production of ion selective electrodes that are soft-bodied and disposable. With features make them especially suitable for clinical, particularly gastroenterological, investigations. We report here on the construction of a combined reference and
M O'Dowd et al.
Journal of biomedical engineering, 10(2), 165-170 (1988-04-01)
Frequent measurement of pH and PCO2 levels of fetal blood during labour provides an insight into the presence and nature of fetal asphyxia, effectiveness of in utero resuscitation and the need for operative delivery. A pH measuring system utilizing a
J M Rawlings et al.
Gut, 26(2), 203-207 (1985-02-01)
Plastic electrodes have been developed for measuring pH in the human gastrointestinal tract. The electrodes have a plastic hydrogen ion sensitive membrane sealed to a length of fluid filled PVC tubing. Two recently developed hydrogen ion sensitive ligands have been
Ying-Jey Guh et al.
Endocrinology, 155(5), 1728-1737 (2014-01-16)
Endothelin-1 (EDN1) is an important regulator of H⁺ secretion in the mammalian kidney. EDN1 enhances renal tubule H⁺-ATPase activity, but the underlying mechanism remains unclear. To further elucidate the role of EDN1 in vertebrates' acid-base regulation, the present study used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service