Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

932191

Sigma-Aldrich

SPhos ChemBeads

Synonym(s):

2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl, S Phos, SPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35O2P
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

P(C4CCCCC4)(C3CCCCC3)c1c(cccc1)c2c(cccc2OC)OC

InChI

1S/C26H35O2P/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21/h9-11,16-21H,3-8,12-15H2,1-2H3

InChI key

VNFWTIYUKDMAOP-UHFFFAOYSA-N

General description

SPhos [2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl] is an air-stable, electron-rich biaryl phosphine ligand developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.

Application

SPhos may be used as a ligand in the following processes:
  • Palladium catalyzed Suzuki-Miyaura cross-coupling reaction between Boc-protected aminomethyltrifluoroborate and aryl chlorides or hetaryl chlorides to form the corresponding aminomethylarenes.
  • Palladium catalyzed Suzuki-Miyaura cross-coupling reaction between 4-methyl-substituted piperidinylzinc reagent and different aryl or heteroaryl iodides to form various substituted piperidines.
  • Intramolecular Suzuki-Miyaura coupling to form the 18-membered macrocyclic ring during the multi-step synthesis of riccardin C.
Utilized in conjunction with palladium to form a highly active catalyst for C-N bond formation.
Highly universal ligand for Suzuki-Miyaura coupling; aryl chlorides, hindered biaryls, heterobiaryls.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

Product is also available as neat ligand (638072)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service