Skip to Content
MilliporeSigma
All Photos(1)

Documents

928305

Sigma-Aldrich

cataCXium® A Pd G3 ChemBeads

Synonym(s):

Mesylate[(di(1-adamantyl)-n-butylphosphine)-2-(2′-amino-1,1′-biphenyl)]palladium(II), [(Di(1-adamantyl)-butylphosphine)-2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, cataCXium-A-Pd-G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H52NO3PPdS
CAS Number:
Molecular Weight:
728.27
UNSPSC Code:
12352101
NACRES:
NA.82

form

solid

Quality Level

composition

~ 4 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

cataCXium® A Pd G3 is a Buchwald third generation precatalyst that can be used in:

  • Direct ortho-arylation of pyridinecarboxylic acids.
  • Catalyzing Suzuki–Miyaura cross-coupling in the synthesis of 1-heteroaryl-3-azabicyclo[3.1.0]hexanes.
  • Palladium-catalyzed carbonylative carboperfluoroalkylation of alkynes.
  • Suzuki–Miyaura coupling reaction of geminal bis(boryl)cyclopropanes in the synthesis of various gem-disubstituted cyclopropanes.
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (761435)

Legal Information

cataCXium is a registered trademark of Umicore AG & Co. KG

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service