Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

927775

Sigma-Aldrich

XPHOS PD G2 ChemBeads

Synonym(s):

2nd Generation XPhos Precatalyst, Chloro(2-dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II), X-Phos aminobiphenyl palladium chloride precatalyst, XPhos-Pd-G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H59ClNPPd
CAS Number:
Molecular Weight:
786.80
UNSPSC Code:
12352101
NACRES:
NA.82

form

solid

Quality Level

composition

~ 5 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

Cl[Pd@@]1(C2=C(C3=C(N1)C=CC=C3)C=CC=C2)P(C4CCCCC4)(C5=CC=CC=C5C6=C(C(C)C)C=C(C(C)C)C=C6C(C)C)C7CCCCC7

InChI

1S/C33H49P.C12H10N.ClH.Pd/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3;1-6,8-9H,13H2;1H;/q;-1;;+2/p-1

InChI key

HLXMDSGYOJBEBA-UHFFFAOYSA-M

General description

XPhos Pd G2 is a bulky monodentate biaryl ligand. It is a Buchwald′s 2nd generation preformed catalyst. It undergoes rapid reductive elimination to form a reactive, monoligated Pd(0) species.

For larger scale uses, product also available in powdered form (741825)

Application

XPhos Pd G2 was employed as catalyst in the following reactions:
  • Palladium-catalyzed Suzuki-Miyaura coupling reactions of potassium organotrifluoroborates and sulfamates.
  • Suzuki-Miyaura cross-coupling reactions of sensitive aryl and heteroarylboronic acids.
  • Synthesis of potassium Boc-protected secondary aminomethyltrifluoroborates, via Suzuki-Miyaura cross-coupling reaction.
2nd Generation Buchwald Precatalysts
  • Widely used for Suzuki-Miyaura cross-coupling reactions
  • ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
    Learn more about ChemBeads products

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service